Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) bromide, anhydrous, 98% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 3115414

Quantity 25g

 View more versions of this product

Catalog No. AA3115414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7789-46-0
98% min.
215.67
Iron(II) bromide, anhydrous
Granular
Br2Fe
MFCD00016081

Specifications

7789-46-0
98% min.
215.67
MFCD00016081
Yellow
Iron(II) bromide, anhydrous
Granular
Br2Fe
Yellow granules
Odorless
25g

Safety and Handling

SDS & Safety Documents

H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 232-168-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit