Alfa Aesar™ Iron(II) bromide, ultra dry, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 4347222

Quantity 100g

 View more versions of this product

Catalog No. AA4347222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-46-0
    Molecular Formula Br2Fe
    Formula Weight 215.67
    MDL Number MFCD00016081
    UN Number UN3260
    Merck Index 14,4042
    Chemical Name or Material Iron (II) bromide, Ultra dry

    Specifications

    Formula Weight 215.67
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    CAS 7789-46-0
    Sensitivity Air Sensitive & Hygroscopic
    Merck Index 14,4042
    Quantity 100g
    Health Hazard 1 Danger
    TSCA TSCA
    Odor Odorless
    Molecular Formula Br2Fe
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Physical Form Powder
    MDL Number MFCD00016081
    UN Number UN3260
    Melting Point 684°C
    Density 4.636g/mL
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Iron (II) bromide, Ultra dry
    Assay 99.995%, (metals basis)

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit