Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) chloride tetrahydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
5kg
Packaging:
Glass bottle
Plastic bottle
13478-10-9
Cl2Fe·4H2O
198.81
WSSMOXHYUFMBLS-UHFFFAOYSA-L
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
16211588
CHEBI:86249
dichloroiron;tetrahydrate
O.O.O.O.Cl[Fe]Cl
13478-10-9
Cl2Fe·4H2O
198.81
WSSMOXHYUFMBLS-UHFFFAOYSA-L
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
16211588
CHEBI:86249
dichloroiron;tetrahydrate
O.O.O.O.Cl[Fe]Cl
Assay Percent Range 99+%
Linear Formula FeCl2·4H2O
Merck Index 14,4043
Arsenic (As) 5ppm max.
Solubility Information Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
Formula Weight 198.81
Physical Form Crystalline Powder, Crystals or Chunks
Percent Purity 99+%
Copper (Cu) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Glass bottle
Solubility Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
Sulfate 200ppm max.
Color Yellow to Green
Melting Point 105.0°C to 110.0°C
Quantity 1kg
Chemical Name or Material Iron(II) chloride tetrahydrate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger