Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) sulfate heptahydrate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
5g
250g
1kg
5kg
Packaging:
Glass Bottle
Plastic bottle
7782-63-0
FeH14O11S
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
7782-63-0
FeH14O11S
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
Assay Percent Range 99.5%
Linear Formula FeSO4·7H2O
Fieser 01,393; 04,237; 05,308
Formula Weight 278.01
Physical Form Crystals
Percent Purity ≥99.5%
Grade Analysis
Chlorine (Cl) 5ppm max.
Copper (Cu) 10ppm max.
Iron (Fe3) 100ppm max.
Lead (Pb) 5ppm max.
Packaging Glass Bottle
Phosphate 10ppm max.
Substances not ppt by NH3 0.05% (as SO4) max.
Total Nitrogen Compounds 0.001% max.
Zinc (Zn) 50ppm max.
Color Blue to Green
Quantity 5g
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less