Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Iron(Ii) Sulfate Heptahydrate, ACS Reagent, Honeywell Fluka™

Click to view available options
Quantity:
5g
250g
1kg
2.5kg
6 x 1kg
7782-63-0
FeH14O11S
278.006
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
7782-63-0
FeH14O11S
278.006
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula FeSO4 · 7H2O
Formula Weight 278.01g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density 1.898g/mL at 25°C
Vapor Pressure 14.6mmHg (25°C)
Quantity 5g
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate

P301 + P312 + P330-P305 + P351 + P338

H302-H315-H319

EINECSNumber : 231-753-5

RTECSNumber : NO8510000