Alfa Aesar™ Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1296706

Quantity 5g

 View more versions of this product

Catalog No. AA1296706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-63-0
    Linear Formula FeSO4·7H2O
    Formula Weight 278.02 (151.91 Anhydrous)
    MDL Number MFCD00149719
    Merck Index 14,4057
    Chemical Name or Material Iron(II) sulfate heptahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 278.02 (151.91 Anhydrous)
    Melting Point 64°C (in -6H2O)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P321-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,4057
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    RTECS Number NO8510000
    TSCA TSCA
    Physical Form Crystalline
    Health Hazard 1 Warning
    Density 1.897g/mL
    Sensitivity Air Sensitive
    MDL Number MFCD00149719
    CAS 7782-63-0
    Linear Formula FeSO4·7H2O
    Color Pale green
    Chemical Name or Material Iron(II) sulfate heptahydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P321-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit