Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Iron(Ii) Sulfate Hydrate, Honeywell Fluka™

Click to view available options
Quantity:
1kg
50kg
6 x 1kg
13463-43-9
FeH2O5S
169.916
MFCD00149722
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
ferrous sulfate monohydrate,iron 2+ sulfate monohydrate,ferrous sulfate hydrate,iron sulfate monohydrate,unii-rib00980vw,iron ii sulfate, monohydrate,iron ii sulfate hydrate,sulfuric acid, iron 2+ salt 1:1 , monohydrate,feroritard,feromax
62712
CHEBI:75834
iron(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Fe+2]
13463-43-9
FeH2O5S
169.916
MFCD00149722
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
ferrous sulfate monohydrate,iron 2+ sulfate monohydrate,ferrous sulfate hydrate,iron sulfate monohydrate,unii-rib00980vw,iron ii sulfate, monohydrate,iron ii sulfate hydrate,sulfuric acid, iron 2+ salt 1:1 , monohydrate,feroritard,feromax
62712
CHEBI:75834
iron(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Fe+2]

Puriss., meets analytical specification of BP, European Pharmacopoeia, exsiccated, 86.0-89.0% FeSO4 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula FeSO4 · aq
Formula Weight 151.91g/mol (anhydrous basis)
Grade puriss.
Concentration 86 to 89.0% FeSO4 basis
Physical State Exsiccated
Quantity 1kg
Chemical Name or Material Iron(Ii) Sulfate Hydrate, Meets analytical specification of Ph. Eur.BP

P273-P305 + P351 + P338-P501

H302-H315-H319-H410

EINECSNumber : 231-753-5