Alfa Aesar™ Iron(II) titanium oxide, 99.8+% (metals basis)

Manufacturer: Alfa Aesar™ 1313730

Quantity 250g

 View more versions of this product

Catalog No. AA1313730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12022-71-8
    Formula Weight 151.75
    MDL Number MFCD00064704
    Synonym Iron(II) titanate
    Chemical Name or Material Iron(II) titanium oxide

    Specifications

    Formula Weight 151.75
    TSCA TSCA
    Synonym Iron(II) titanate
    Mesh Size −100 Mesh
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00064704
    Quantity 250g
    CAS 12022-71-8
    Chemical Name or Material Iron(II) titanium oxide
    Assay ≥99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit