Alfa Aesar™ Iron(III) 1,3-diphenyl-1,3-propanedionate

Manufacturer: Alfa Aesar™ H3227806

Quantity 5g

 View more versions of this product

Catalog No. AAH3227806

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C45H33FeO6
    Formula Weight 725.61
    MDL Number MFCD14636458
    Synonym Iron(III) (DBM)3; Tris(dibenzoylmethanato)iron(III)
    Chemical Name or Material Iron(III) 1,3-diphenyl-1,3-propanedionate

    Specifications

    Formula Weight 725.61
    Molecular Formula C45H33FeO6
    MDL Number MFCD14636458
    Synonym Iron(III) (DBM)3; Tris(dibenzoylmethanato)iron(III)
    Quantity 5g
    Chemical Name or Material Iron(III) 1,3-diphenyl-1,3-propanedionate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit