Alfa Aesar™ Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%

Manufacturer: Alfa Aesar™ 8930730

Quantity 250g

 View more versions of this product

Catalog No. AA8930730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7321-53-1
    Molecular Formula C24H45FeO6
    Formula Weight 485.47
    MDL Number MFCD00014006
    UN Number UN1268
    Chemical Name or Material Iron (III) 2-ethylhexanoate, Nominally 50% in mineral spirits

    Specifications

    Formula Weight 485.47
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Concentration Fe 6%
    UN Number UN1268
    Physical Form Liquid
    Density 0.91g/mL
    MDL Number MFCD00014006
    Health Hazard 2 GHS H Statement
    H340-H350-H372-H304-H226
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
    Molecular Formula C24H45FeO6
    CAS 7321-53-1
    Health Hazard 1 Danger
    Color Brown
    Quantity 250g
    Chemical Name or Material Iron (III) 2-ethylhexanoate, Nominally 50% in mineral spirits

    GHS H Statement
    H340-H350-H372-H304-H226
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit