Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) bromide, 90+%, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
10031-26-2
Br3Fe
295.56
MFCD00016082
10031-26-2
Br3Fe
295.56
MFCD00016082
Lewis acid
Assay Percent Range 90% min. (Iodometry)
Linear Formula FeBr3
Merck Index 15, 4045
Solubility Information Solubility in water: soluble
Formula Weight 295.56
Physical Form Powder
Percent Purity 90+%
Packaging Glass bottle
Color Beige-Yellow to White
Melting Point 200°C
Quantity 25g
Chemical Name or Material Iron(III) bromide
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 233-089-1