Alfa Aesar™ Iron(III) bromide, anhydrous, 98+%

Manufacturer: Alfa Aesar™ 3112506

Quantity 5g

 View more versions of this product

Catalog No. AA3112506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10031-26-2
    Formula Weight 295.57
    MDL Number MFCD00016082
    Merck Index 14,4018
    Chemical Name or Material Iron(III) bromide, Anhydrous

    Specifications

    Formula Weight 295.57
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Odor Odorless
    Physical Form Crystalline
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H302-H335-H319
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes serious eye irritation.
    Sensitivity Light Sensitive & Hygroscopic
    CAS 10031-26-2
    TSCA TSCA
    MDL Number MFCD00016082
    Merck Index 14,4018
    Color Dark red
    Chemical Name or Material Iron(III) bromide, Anhydrous
    Assay ≥98%

    GHS H Statement
    H314-H302-H335-H319
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit