Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) chloride hexahydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
10025-77-1
Cl3Fe·6H2O
270.29
Ferric chloride hexahydrate
10025-77-1
Cl3Fe·6H2O
270.29
Ferric chloride hexahydrate
Lewis acid
Assay Percent Range 99+%
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15,4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol,acetone and ether
Formula Weight 270.29
Physical Form Lumps
Percent Purity ≥99%
Grade Extra Pure
Ferrous Iron (Fe II) 500ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
Sulfate 100ppm max.
Boiling Point 280.0°C to 285.0°C
Color Brown to Yellow
Melting Point 37.0°C
Quantity 1kg
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less