Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Iii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™

Click to view available options
Quantity:
5g
100g
500g
6 x 500g
10025-77-1
MFCD00149712
10025-77-1
MFCD00149712

ACS Reagent, 97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula FeCl3 · 6H2O
UN Number 3260
Formula Weight 270.3g/mol
Percent Purity 97%
Grade ACS Reagent
Vapor Pressure 1mmHg (194°C)
Quantity 5g
Chemical Name or Material Iron(Iii) Chloride Hexahydrate

P280-P305 + P351 + P338

H290-H302-H315-H318

EINECSNumber : 231-729-4

RTECSNumber : NO5425000