Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) chloride solution, Purum, 45% FeCl3 basis, Honeywell Fluka™

Catalog Number 6000840
Click to view available options
Quantity:
2.5 L
7705-08-0
Cl3Fe
162.20
MFCD00011005
RBTARNINKXHZNM-UHFFFAOYSA-K
iron(3+) trichloride
[Cl-].[Cl-].[Cl-].[Fe+3]
7705-08-0
Cl3Fe
162.20
MFCD00011005
RBTARNINKXHZNM-UHFFFAOYSA-K
iron(3+) trichloride
[Cl-].[Cl-].[Cl-].[Fe+3]

Purum, 45% FeCl3 basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Quantity 2.5 L
Linear Formula FeCl3
UN Number 2582
Formula Weight 162.2g/mol
Grade purum
Concentration or Composition (by Analyte or Components) 45% FeCl3 basis
Chemical Name or Material Iron(Iii) Chloride Solution
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H290-H315-H318
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit