Alfa Aesar™ Iron(III) ethoxide, soluble polymeric mixture

Manufacturer: Alfa Aesar™ 4586104

Quantity 2g

 View more versions of this product

Catalog No. AA4586104

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00078028
    UN Number UN2926
    Chemical Name or Material Iron (III) ethoxide, Soluble polymeric mixture

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00078028
    UN Number UN2926
    Color Brown
    Quantity 2g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
    Physical Form Solid
    Solubility Information Soluble in hexane and ethanol
    Chemical Name or Material Iron (III) ethoxide, Soluble polymeric mixture

    GHS H Statement
    H314-H318-H228
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable solid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit