Alfa Aesar™ Iron(III) hydroxide, 8.0% in Kensol 50H, nanoparticle dispersion, <5.0nm APS Number Weighted

Manufacturer: Alfa Aesar™ 4724236

Quantity 500g

 View more versions of this product

Catalog No. AA4724236

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 20344-49-4
    Linear Formula FeO(OH)
    Formula Weight 88.85
    MDL Number MFCD00064782
    Merck Index 14,4025
    Synonym Iron oxyhydroxide in Kensol 50H
    Chemical Name or Material Iron(III) hydroxide, Nanoparticle dispersion

    Specifications

    Formula Weight 88.85
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00064782
    Synonym Iron oxyhydroxide in Kensol 50H
    Color Red-brown
    Health Hazard 2 GHS H Statement
    H350
    May cause cancer.
    Density 0.88g/mL
    CAS 20344-49-4
    Merck Index 14,4025
    TSCA TSCA
    Particle Size <5.0nm APS Number Weighted
    Physical Form Liquid
    Health Hazard 1 Danger
    Linear Formula FeO(OH)
    Quantity 500g
    Concentration 8.0% in Kensol 50H
    Chemical Name or Material Iron(III) hydroxide, Nanoparticle dispersion

    GHS H Statement
    H350
    May cause cancer.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit