Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iron(III) isopropoxide, 98%

Manufacturer:  Alfa Aesar™ 4487306

Quantity 5g

 View more versions of this product

Catalog No. AA4487306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14995-22-3
Fe[OCH(CH3)2]3
MFCD00070436
Iron triisopropoxide
C9H21FeO3
233.11
UN3181
Iron (III) isopropoxide

Specifications

14995-22-3
Solid Mass
Fe[OCH(CH3)2]3
MFCD00070436
Dark gray
5g
Iron (III) isopropoxide
Ampule
C9H21FeO3
233.11
UN3181
Iron triisopropoxide
Moisture Sensitive
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

storageNote1 :

  • Ampouled under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit