Alfa Aesar™ Iron(III) meso-tetraphenylporphine chloride

Manufacturer: Alfa Aesar™ 1882903

Quantity 1g

 View more versions of this product

Catalog No. AA1882903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16456-81-8
    Formula Weight 704.04
    MDL Number MFCD00012152
    Synonym 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride
    Chemical Name or Material Iron(III) meso-tetraphenylporphine chloride

    Specifications

    Formula Weight 704.04
    MDL Number MFCD00012152
    Physical Form Crystalline
    CAS 16456-81-8
    Synonym 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride
    Quantity 1g
    Chemical Name or Material Iron(III) meso-tetraphenylporphine chloride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit