Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis), Acros Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
7782-61-8
FeN3O9·9H2O
404
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate,iron iii nitrate nonahydrate,unii-vmx4uop3vn,ferric nitrate, nonahydrate,iron iii nitrate,vmx4uop3vn,iron, reference standard solution,ferricnitratenonahydrate,nitric acid, iron 3+ salt, nonahydrate,iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
7782-61-8
FeN3O9·9H2O
404
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate,iron iii nitrate nonahydrate,unii-vmx4uop3vn,ferric nitrate, nonahydrate,iron iii nitrate,vmx4uop3vn,iron, reference standard solution,ferricnitratenonahydrate,nitric acid, iron 3+ salt, nonahydrate,iron iii nitrate hydrate
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
For ultra trace analysis
Assay Percent Range 13.5 to 14.2% (Fe) (Iodometry)
Linear Formula Fe(NO3)3·9H2O
Merck Index 15,4054
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,acetone
Formula Weight 404
Physical Form Crystals
Percent Purity 99.9995%
Grade Trace Metal Basis
Density 1.6800g/mL
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone
Specific Gravity 1.68
Total Trace Metal Impurities 5ppm max.
Color Purple to White or Violet
Melting Point 47.0°C
Quantity 25g
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti

GHS Signal Word: Warning