Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) oxide, 95%, pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
25kg
1309-37-1
Fe2O3
159.69
Ferric oxide
1309-37-1
Fe2O3
159.69
Ferric oxide
Assay Percent Range 94% min. (Iodometry)
Linear Formula Fe2O3
Ash 0.3% max. (1000°C, 0.5 h)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether
Formula Weight 159.69
Percent Purity 95%
Grade Pure
Density 5.2400g/mL
Loss on Drying 0.3% max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether
Specific Gravity 5.24
Melting Point 1538.0°C
Quantity 25kg
Chemical Name or Material Iron(III) oxide
Show More Show Less

EINECSNumber : 215-168-2