Iron(III) oxide, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1259322

Quantity 100g

 View more versions of this product

Catalog No. AA1259322

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1309-37-1
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula Fe2O3
    Formula Weight 159.69
    MDL Number MFCD00011008
    Chemical Name or Material Iron(III) oxide

    Specifications

    Formula Weight 159.69
    Quantity 100g
    Color Red-brown
    Density 5.24g/cm3
    EINECS Number 215-168-2
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula Fe2O3
    Appearance Red-brown powder
    MDL Number MFCD00011008
    CAS 1309-37-1
    Health Hazard 1 H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Iron(III) oxide

    H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit