Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass Bottle
1309-37-1
Fe2O3
159.69
Ferric oxide
1309-37-1
Fe2O3
159.69
Ferric oxide
for ultra trace analysis
Assay Percent Range 99.999%
Linear Formula Fe2O3
Fieser 17,158
Merck Index 15,4055
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids,soluble in alcohol and ether
Formula Weight 159.69
Physical Form Powder
Percent Purity 99.999%
Grade Trace Metal Basis
Density 5.2400g/mL
Packaging Glass Bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in acids, soluble in alcohol and ether
Specific Gravity 5.24
Total Trace Metal Impurities 0.001% max
Color Brown to Brown-Red or Rust
Melting Point 1538.0°C
Quantity 250g
Chemical Name or Material Iron(III) oxide
Show More Show Less

EINECSNumber : 215-168-2