Alfa Aesar™ Iron(III) oxide, nanopowder, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 44119A1

Quantity 1kg

 View more versions of this product

Catalog No. AA44119A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1309-37-1
    Molecular Formula Fe2O3
    Formula Weight 159.69
    MDL Number MFCD00011008
    Merck Index 14,4028
    Chemical Name or Material Iron (III) oxide

    Specifications

    Formula Weight 159.69
    Physical Form Nanopowder
    Molecular Formula Fe2O3
    Surface Area 250m2/g
    Merck Index 14,4028
    Quantity 1kg
    TSCA TSCA
    MDL Number MFCD00011008
    Color Red-brown
    Melting Point 1565°C
    Density 5.24g/mL
    Odor Odorless
    Particle Size 3nm APS
    CAS 1309-37-1
    RTECS Number NO7400000
    Chemical Name or Material Iron (III) oxide
    Assay 99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit