Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iron(III) oxide, nanopowder, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4411918

Quantity 50g

 View more versions of this product

Catalog No. AA4411918


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1309-37-1
159.69
14,4028
Fe2O3
MFCD00011008
Iron (III) oxide

Specifications

1309-37-1
Fe2O3
MFCD00011008
5.24g/mL
3nm APS
Red-brown
50g
99.95%, (metals basis)
Nanopowder
159.69
14,4028
Odorless
250m2/g
1565°C
Iron (III) oxide

Safety and Handling

SDS & Safety Documents

RTECSNumber : NO7400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit