Iron(III) sulfate hydrate, Reagent Grade, Alfa Aesar™

Manufacturer: Alfa Aesar 3331630

Quantity 250g

 View more versions of this product

Catalog No. AA3331630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15244-10-7
    Physical Form Granular
    Molecular Formula Fe2(SO4)3·xH2O
    Formula Weight 399.87 (Anhydrous)
    MDL Number MFCD00011007
    Chemical Name or Material Iron(III) sulfate hydrate
    Grade Reagent Grade

    Specifications

    Formula Weight 399.87 (Anhydrous)
    Quantity 250g
    Color Gray-white
    EINECS Number 233-072-9
    Physical Form Granular
    Odor Odorless
    Molecular Formula Fe2(SO4)3·xH2O
    Appearance Grayish-white powder or granules
    MDL Number MFCD00011007
    CAS 15244-10-7
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Iron(III) sulfate hydrate
    Grade Reagent Grade

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit