Alfa Aesar™ Iron naphthenate, 80% in mineral spirits (12% Fe)

Manufacturer: Alfa Aesar™ 3859036

Quantity 500g

 View more versions of this product

Catalog No. AA3859036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1338-14-3
    MDL Number MFCD00147295
    UN Number UN1993
    Chemical Name or Material Iron naphthenate

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00147295
    UN Number UN1993
    Color Brown
    Quantity 500g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Density 1.05g/mL
    CAS 1338-14-3
    Concentration 80% in min.eral spirits (12% Fe)
    Flash Point >38° (100°F)
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H340-H350-H372-H304-H226
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    Chemical Name or Material Iron naphthenate

    GHS H Statement
    H340-H350-H372-H304-H226
    May cause genetic defects.
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit