Iron Nickel powder, greater-than99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3668314

Quantity 25g

 View more versions of this product

Catalog No. AA3668314

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Powder
    Assay Percent Range >99.5% (metals basis)
    Molecular Formula Ni:Fe; 50:50 wt%
    MDL Number MFCD01741928
    Chemical Name or Material Iron Nickel powder

    Specifications

    Quantity 25g
    Color Silver-gray
    DOT Information DOT Class: 4.1; Packing Group: II
    Health Hazard 2 Keep container tightly closed. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
    Physical Form Powder
    Assay Percent Range >99.5% (metals basis)
    Molecular Formula Ni:Fe; 50:50 wt%
    MDL Number MFCD01741928
    Odor Odorless
    Health Hazard 1 H228-H351-H319-H317-H335: Flammable solid. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation.
    Appearance Silver gray powder
    Chemical Name or Material Iron Nickel powder

    Keep container tightly closed. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.

    H228-H351-H319-H317-H335: Flammable solid. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit