Learn More
Sigma Aldrich Iron oxide (II,III), nanoparticles
Catalog No. 9001481ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
90-014-81ML 1 mL
1 options
Catalog No. 90-014-81ML Supplier Sigma Aldrich Supplier No. 9001481ML
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Recent docking studies highlight the potential interaction of iron oxide (II,III) nanoparticles (IONPs) with the SARS-CoV-2 spike protein receptor binding domain. This may target viral entry in host cells and also render virions inactive.Iron oxide nanoparticles exhibit biocompatibility and non-toxicity and are used as magnetic resonance imaging contrast agents, gene carriers for gene therapy, therapeutic agents for hyperthermia-based cancer treatment, magnetic sensing probes for in vivo diagnostics and for target specific drug delivery.Product of Ocean Nanotech, LLC.
Specifications
Quantity 1 mL