Iron pieces, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4509036

Quantity 500g

 View more versions of this product

Catalog No. AA4509036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    Physical Form Irregular Shape, 0.125-0.25 In.
    Assay Percent Range 99.99% (metals basis)
    MDL Number MFCD00010999
    Chemical Name or Material Iron pieces

    Specifications

    Physical Form Irregular Shape, 0.125-0.25 In.
    Quantity 500g
    EINECS Number 231-096-4
    Assay Percent Range 99.99% (metals basis)
    MDL Number MFCD00010999
    CAS 7439-89-6
    Chemical Name or Material Iron pieces

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit