Iron powder, 99.5%, Alfa Aesar™

Apparent density 2.0-3.0g/cm3

Manufacturer: Alfa Aesar 1021436

Quantity 500g

 View more versions of this product

Catalog No. AA1021436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    Physical Form Spherical Powder
    Assay Percent Range 99.5% (Metals basis)
    Molecular Formula Fe
    MDL Number MFCD00010999
    Chemical Name or Material Iron, Approximately 98% spherical, 2% fragments

    Specifications

    Physical Form Spherical Powder
    Quantity 500g
    Color Gray
    DOT Information DOT Class: 4.1, Packaging Group: II
    EINECS Number 231-096-4
    Assay Percent Range 99.5% (Metals basis)
    Molecular Formula Fe
    MDL Number MFCD00010999
    CAS 7439-89-6
    Health Hazard 1 H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.
    Appearance Gray powder
    Chemical Name or Material Iron, Approximately 98% spherical, 2% fragments

    H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit