Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron powder, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1062114

Quantity 25g

 View more versions of this product

Catalog No. AA1062114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7439-89-6
99.998% (Metals basis)
MFCD00010999
Powder
Fe
Iron

Specifications

25g
Gray powder
Powder
Fe
Gray
7439-89-6
7.87g/cm3 at 20°C
99.998% (Metals basis)
MFCD00010999
Iron

Safety and Handling

SDS & Safety Documents

H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-096-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit