Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly Bottle
7782-61-8,7697-37-2,7732-18-5
FeH18N3O18
403.992
MFCD00010999
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]
7782-61-8,7697-37-2,7732-18-5
FeH18N3O18
403.992
MFCD00010999
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
iron(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Fe+3]

For atomic absorption. (1mL = 1mg Fe) Solute: Ferric Nitrate Solvent: Dilute Nitric Acid

Concentration 990 to 1010 ppm
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Physical Form Liquid
Grade Certified
Packaging Poly Bottle
Traceability to NIST LOT 3126a Iron reference std.
Vapor Pressure 14mmHg
pH 1
Melting Point 0°C
Boiling Point 100°C
Color Undesignated
Quantity 500mL
Chemical Name or Material Iron, Reference Standard Solution, 1000ppm ±1%
Show More Show Less

DOTInformation : DOT Class 8, : Corrosive