Alfa Aesar™ Iron rod, 12mm (0.47 in.) dia., Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 11500DO

Quantity 150mm

 View more versions of this product

Catalog No. AA11500DO

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    MDL Number MFCD00010999
    Chemical Name or Material Iron rod, 12mm (0.47 in.) dia.
    Grade Puratronic™

    Specifications

    MDL Number MFCD00010999
    CAS 7439-89-6
    Weight ≈0.89g/mm
    Quantity 150mm
    TSCA TSCA
    Physical Form Rod
    Chemical Name or Material Iron rod, 12mm (0.47 in.) dia.
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit