Alfa Aesar™ Iron slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.95+% (metals basis)

Manufacturer: Alfa Aesar™ 0076918

Quantity 50g

 View more versions of this product

Catalog No. AA0076918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    MDL Number MFCD00010999
    Chemical Name or Material Iron slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length

    Specifications

    Physical Form Slug
    MDL Number MFCD00010999
    TSCA TSCA
    CAS 7439-89-6
    Quantity 50g
    Chemical Name or Material Iron slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length
    Assay ≥99.95%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit