Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Standard, 1mL = 1mg Fe, 1000ppm Fe, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 420016

Packaging Poly natural
Quantity 500mL

 View more versions of this product

Catalog No. 420016

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Chemical Identifiers

10025-77-1 (Ferric Chloride Hexahydrate),7647-01-0 (Hydrochloric Acid),7732-18-5 (Water, Deionized)
Liquid
Iron Standard

Specifications

10025-77-1 (Ferric Chloride Hexahydrate),7647-01-0 (Hydrochloric Acid),7732-18-5 (Water, Deionized)
Poly natural
1 mL = 1 mg Fe
500mL
Liquid
1000ppm Fe
Clear, light to dark amber
Iron Standard
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit