Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron wire, 99.99% (metals basis), Alfa Aesar™
Click to view available options
Quantity:
1m
5m
Specifications
Quantity | 1m |
Assay Percent Range | 99.99% (metals basis) |
Physical Form | ∽1.5g/m |
Chemical Name or Material | Iron wire |
Safety and Handling
H500
EINECSNumber : 231-096-4
TSCA : Yes
Recommended Storage : Ambient temperatures