Alfa Aesar™ Iron wire,1mm dia., 99.995% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar™ 10935BW

Quantity 50cm

 View more versions of this product

Catalog No. AA10935BW

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-89-6
    Molecular Formula Fe
    MDL Number MFCD00010999
    Chemical Name or Material Iron

    Specifications

    Quantity 50cm
    Density 7.87g/cm3 at 20°C
    EINECS Number 231-096-4
    Physical Form Wire, 0.25mm (0.01 in.) dia.
    Molecular Formula Fe
    Appearance Gray wire
    Assay Percent Range 99.995% (Metals basis)
    MDL Number MFCD00010999
    CAS 7439-89-6
    Chemical Name or Material Iron

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit