Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micronova™ Irradiated Plastic ACME Threaded Handles

A series of irradiated, plastic coated fiberglass, fixed length and extendable handles. Designed with ACME Connect threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.

$129.00 - $554.00

Specifications

For Use With (Application) Mop Handle for Cleaning and Sanitizing Cleanrooms
Autoclavable Autoclavable
Color White
Disposable No
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Length (Metric) Length (English) Type Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Length (English) Type Price Quantity & Availability  
19-166-383
View Documents
Micronova™
STAH12WHIR
30.5 cm 12 in. Fixed
Case of 6 for $129.00
Add to cart
 
19-166-384
View Documents
Micronova™
STAEH036WHIR
46 to 91 cm 18 to 36 in. Extendible
Case of 6 for $554.00
Add to cart
 
Description

Description

The Micronova Irradiated Plastic ACME Threaded Handles are made from lightweight fiberglass coated in plastic and are available in a range of extendable and fixed length options. The ACME Connect threaded ends attach directly to the plastic (QP series) adapters, or use the SSA-2 ACME Thread Adapters to attach to Quick Disconnect (QD series) mop heads.

  • Irradiated, lightweight, plastic coated fiberglass handles
  • Perfect match for irradiated mop frames
  • ACME Connect threaded end connection
  • Available fixed length or extendable
    • Fixed lengths range from 12 to 60 inches (30 cm to 1.5 m)
    • Extendable maximum lengths range from 36 in to 11.5 ft (90 cm to 3.5 m)
  • ACME Connect threaded end attaches directly to the plastic (QP series) adapters
  • Attach to (QD series) mop heads via the SSA-2 ACME Thread Adapters
  • Gamma Irradiated for aseptic environments
  • Autoclavable
Specifications

Specifications

Mop Handle for Cleaning and Sanitizing Cleanrooms
White
Non-sterile
6/Cs.
Autoclavable
No
Plastic Coated Fiberglass, Plastic Fittings
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit