Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI3G10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271049JF646

Catalog No. N271049J646


Add to cart

Description

Description

IRS1 Monoclonal antibody specifically detects IRS1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

IRS1
OTI3G10
Immunohistochemistry
Affinity Purified, Conjugated
IRS1
Recombinant protein fragment corresponding to amino acids 1-280 of human IRS1 (NP_005535) produced in E.coli.
Immunogen affinity purified
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction
Primary
3667.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
HIRS-1, insulin receptor substrate 1, IRS-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit