Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irsogladine maleate, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
84504-69-8
C13H11Cl2N5O4
372.16
MFCD00873566
PJLVTVAIERNDEQ-BTJKTKAUSA-N
irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234
5282435
(2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl
This item is not returnable. View return policy
84504-69-8
C13H11Cl2N5O4
372.16
MFCD00873566
PJLVTVAIERNDEQ-BTJKTKAUSA-N
irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234
5282435
(2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl

PDE4 inhibitor; antiulcer agent

Phosphodiesterase 4 (PDE4) inhibitor that displays gastroprotective properties. Prevents gastric mucosal injury induced by monochloramine and ischemia-reperfusion. Activates gap-junctional intercellular communication improving gastric mucosal barrier function. Inhibits superoxide production in human neutrophils and inhibits in vitro and in vivo angiogenesis.
Quantity 50mg
Formula Weight 372.18
Percent Purity >99%
Chemical Name or Material Irsogladine maleate

Recommended Storage : Desiccate at Room Temperature