Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Air Boss™ Defense IRT Hazmat Berms with Carry Bag

No tools required

Specifications

Color Black
Product Type Hazmat berm
For Use With (Application) Seating and sealing the floor area within decon shelters
Includes Carry bag
Material Polyester, polypropylene and stainless steel connectors
View More Specs

Products 4
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity & Availability  
19-130-0630
View Documents
Air Boss™ Defense
HBR4545
45 in. 45 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0631
View Documents
Air Boss™ Defense
HBR5555
55 in. 55 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0632
View Documents
Air Boss™ Defense
HBR7296
96 in. 72 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-0633
View Documents
Air Boss™ Defense
HBR9696
96 in. 96 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Optimized pre-fabricated design for rapid deployment
  • Polypropylene and stainless-steel connectors
  • Compact and lightweight for portability
  • Fully collapsible and foldable
  • Less storage space required
  • Non-corrosive

Specifications

Specifications

Black
Seating and sealing the floor area within decon shelters
Polyester, polypropylene and stainless steel connectors
Hazmat berm
Carry bag
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit