Learn More
IRT Hazmat Berms with Carry Bag
No tools required
Manufacturer:  TVI CorporationSupplier Diversity Partner HBR5555
 View more versions of this product
Catalog No. 19-130-0631
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyester, polypropylene and stainless steel connectors
Black
55 in.
Hazmat berm
Seating and sealing the floor area within decon shelters
Carry bag
55 in.