Learn More
Air Boss™ Defense IRT Individual Decon Shelter
Manufacturer:  Air Boss™ Defense SD1TTB02GZ
Catalog No. 19-065-289
Add to cart
Description
Description
Effectively handles decontamination of a single ambulatory casualty. Plus, shelter's tall profile is suitable for decon of personnel in full PPE.
Specifications
Specifications
Safety
Blue
152.4cm
60 in.
55 lb.
182.8cm
60 x 72 in. (152.4 x 182.8cm)
7 Total 1 gpm standard body spray, 16 in. dia. Snorkelled port per side, interior berm, 2-panel zippered end doors with vel-cro strip, anchor kit, repair kit, hose adaptor kit, transport bag
152.4cm
60 in.
Decontamination shelter
24.94kg
72 in.
Documents
Accessories