IRT ISO-POD™ Negative Pressure Individual Isolation System
For safe, temporary patient isolation, holding and transport
Manufacturer: TVI CorporationSupplier Diversity Partner PODA4ECO
 View more versions of this product
Catalog No. 19-150-1140
  Check Availability
Add to Cart
    Specifications
    Product Type Economy solation system
    Case Material PVC
    Includes Two HEPA (in)/one OVAG (out) canister, rechargeable 6V NiMH 4 Hours battery and charger, 1 window/side, 2 pairs Nitrile gloves, 1 snorkel
  • Description & Specifications
Related Products
Product Content Feedback