Learn More
Air Boss Defense IRT Outdoor Negative Pressure Isolation Shelters
Isolation chamber ensures state-of-the-art performance, reliability and rapid deployment
$25400.00
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity & Availability | |||||
19-130-2239
|
Air Boss Defense
SISWXA10UZ |
288 x 192 x 108 in. (731.5 x 487.6 x 274.32cm) |
Each for $25,400.00
|
|
Description
Corrosion-resistant anodized aluminum frame with chemical-resistant fabric provides for long life and reliability. Shelters sized for three or eight gurneys. Can also be used for positive pressure.
- Integral zippered 5 ft. airlock with full airlock functionality
- Sidewall and overhead attachment points for monitoring equipment
- Thermal screen for reduced shelter heat gain
- Snorkeled ductwork for filtration unit
- Quick disconnect for chamber air pressure feedback tube
- Standard windows on each side and double end viewing portals
- Up to two utility port, two viewing and two pass-thru interchangeable windows, one airlock door and two end doors, and seven lights
Cat. No. 19-130-0297 has full-length windows on either side.
Specifications
Isolation shelter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.