Learn More
Air Boss™ Defense IRT Two-Line Decontamination Shelters
Manufacturer:  Air Boss™ Defense SD2TZB08GZ
 View more versions of this product
Catalog No. 19-065-287
Add to cart
Description
Description
Accommodate two lines of ambulatory casualties or one line of nonambulatory casualties, all within a compact footprint.
Specifications
Specifications
Safety
274.32cm
108 in.
240 in.
145 lb.
243.84cm
240 x 96 x 108 in. (609.6 x 243.84 x 274.32cm)
20 body spray, 1 Center divider, 6 isolation curtains with tie down, active curtains, 2 Water booms and nozzle, interior catch basin, 2 zippered Clothing ports/side, 2 6 in. dia. snorkelled ports/side, 1 14 in. dia. snorkelled ports/Side, 3-p
Blue
609.6 cm
2 Line decontamination shelter
65.77kg
96 in.
Documents
Accessories