IRT Water Storage Bladders
Effectively collect wastewater runoff from IRT decontamination shelters
Manufacturer: TVI CorporationSupplier Diversity Partner WSB1000G
 View more versions of this product
Catalog No. 19-130-0637
  Check Availability
Add to Cart
    Specifications
    Includes Protective ground Cloth and transport bag, integral pressure relief valve, ball valve, back flow prevention valve and cam-lok
    Product Type Waste storage bladder
    Material Polyester
    For Use With (Application) Collects waste Water run-off from IRT decontamination shelters
    Capacity (English) 1000 gal.
    Capacity (Metric) 3785.4L
  • Description & Specifications
Related Products
Product Content Feedback