Learn More
ORS Nasco Irwin™ Vise-Grip™ Straight Jaw Locking Pliers
Provide maximum contact on flat, square, or hex work
Manufacturer:  ORS Nasco 58610R3
Catalog No. 19-313-658
Add to cart
Description
Description
  • Turn screw to adjust pressure and fit work; stays adjusted for repetitive use
  • Constructed of high-grade heat-treated alloy steel for maximum toughness and durability
  • Guarded release trigger quickly unlocks and protects from accidental release
  • Hardened teeth are designed to grip from any angle
  • For tightening, clamping, twisting and turning
Specifications
Specifications
10 in.
Alloy Steel
Straight
1.31 lb.
25.4 cm
Adjustable mechanical locking plier
0 to 1.6 in.