Learn More
Aviva Systems Biology Corp IRX1 Antibody - middle region (ARP47681_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47681_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IRX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IRX1. Alternate names: IRX-5, IRXA1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59636